Please login to your online account to display your discounted pricing

anti-Muscleblind-like 1, Clone: HL-1822 3A4-1E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11037256SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11037256T

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Clone HL-1822 3A4-1E9
Antigen Muscleblind-like 1
Isotype IgG1, κ
Gene Accession No. Q9NR56
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Format Supernatant
Immunogen Recombinant human Muscleblind-like 1 protein. [UniProt No. Q9NR56]
Host Species Mouse
Quantity 0.1mL
Conjugate Unlabeled
Gene ID (Entrez) 4154