Please login to your online account to display your discounted pricing

anti-MUL1, Clone: 5H6.2D5, Novus Biologicals™

$109.00 - $259.02

Specifications

Clone 5H6.2D5
Host Species Mouse
Isotype κ
Immunogen Partial recombinant protein made to a C-terminal portion of the human MUL1 protein (between residues 200-352) [UniProt Q969V5]
Antigen MUL1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP231361

 
novus biologicals canada
NBP231361
Unlabeled 0.1mg Each for $259.02

NBP231361SS

 
novus biologicals canada
NBP231361SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 5H6.2D5
Host Species Mouse
Isotype κ
Immunogen Partial recombinant protein made to a C-terminal portion of the human MUL1 protein (between residues 200-352) [UniProt Q969V5]
Antigen MUL1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human. The immunogen sequence shows 99% similarity to Monkey's MUL1 and is 84% similar to isoform 1 and 3 of Mouse's MUL1 .
Gene Accession No. Q969V5
Gene ID (Entrez) 79594
Formulation PBS
Concentration 1.0mg/mL