Please login to your online account to display your discounted pricing

anti-MUC-1, Clone: E29, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2294011ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2294011M

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Formulation Culture Medium
Test Specificity In Western blotting, it recognizes proteins in MW range of 265-400kDa, identified as different glycoforms of EMA. This protein is proteolytically cleaved into alpha and beta subunits that form a heterodimeric complex. The N-terminal alpha subunit and the C-terminal beta subunit. The alpha subunit has cell adhesive properties. Can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The beta subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinoma tissues. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.
Primary or Secondary Primary
Format Concentrated
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Quantity 1mL
Isotype λ
Regulatory Status RUO
Gene ID (Entrez) 4582
Cross Reactivity Human
Gene Accession No. P15941
Conjugate Unlabeled
Immunogen A delipidated extract of human milk fat globule membranes was used as the immunogen for the MUC-1 (EMA) E29 antibody.
Clone E29
Antigen MUC-1