Please login to your online account to display your discounted pricing

anti-mtTFA, Clone: 18G102B2E11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171648

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171648

  • / Each
 

Description & Specifications

Specifications

Antigen mtTFA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Clone 18G102B2E11
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Rat
Format Purified
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. Q00059
Host Species Murine
Immunogen Human mtTFA [Swiss-Prot No. Q00059]
Isotype κ
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 7019
Target Species Human
Target Species Rat