Please login to your online account to display your discounted pricing

anti-MS4A1/CD20, Clone: B9E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329271ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329271M

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Format Purified
Gene Accession No. P11836
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Regulatory Status RUO
Concentration 0.2mg/mL
Isotype κ
Primary or Secondary Primary
Gene ID (Entrez) 931
Conjugate Unlabeled
Cross Reactivity Human
Antigen MS4A1/CD20
Host Species Mouse
Clone B9E9
Immunogen Lymphoblastoid cell line Daudi
Quantity 1mL