Please login to your online account to display your discounted pricing

anti-MS4A1/CD20, Clone: B9E9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232927

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232927

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Isotype κ
Clone B9E9
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 931
Concentration 0.2mg/mL
Conjugate Unlabeled
Antigen MS4A1/CD20
Format Purified
Gene Accession No. P11836
Host Species Mouse
Quantity 0.5mL
Immunogen Lymphoblastoid cell line Daudi