Please login to your online account to display your discounted pricing

anti-MS4A1/CD20, Clone: B9E9, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone B9E9
Host Species Mouse
Isotype κ
Immunogen Lymphoblastoid cell line Daudi
Antigen MS4A1/CD20
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232927

 
novus biologicals canada
NBP232927
Unlabeled 0.5mL Each for $321.00

NBP2329271M

 
novus biologicals canada
NBP2329271ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone B9E9
Host Species Mouse
Isotype κ
Immunogen Lymphoblastoid cell line Daudi
Antigen MS4A1/CD20
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P11836
Gene ID (Entrez) 931
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL