Please login to your online account to display your discounted pricing

anti-MRP1, Clone: IU2H10, Novus Biologicals™


Manufacturer: novus biologicals canada  NB4001561ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB4001561ML

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant peptide (a.a. 1-33) of human MRP1. [UniProt No. P33527].
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Concentration 1.0mg/mL
Conjugate Unlabeled
Format Purified
Quantity 1mL
Host Species Mouse
Clone IU2H10
Isotype IgG1
Antigen MRP1
Formulation Tris-glycine, 150mM NaCl
Gene ID (Entrez) 4363
Gene Accession No. P33527