Please login to your online account to display your discounted pricing

anti-Moesin, Clone: MSN491, Novus Biologicals™

$329.00 - $429.00

Specifications

Clone MSN491
Host Species Mouse
Isotype κ
Immunogen Recombinant human Moesin protein
Antigen Moesin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232875

 
novus biologicals canada
NBP232875
Unlabeled 0.5mL Each for $329.00

NBP2328751M

 
novus biologicals canada
NBP2328751ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone MSN491
Host Species Mouse
Isotype κ
Immunogen Recombinant human Moesin protein
Antigen Moesin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Gene Accession No. P26038
Gene ID (Entrez) 4478
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL