Please login to your online account to display your discounted pricing

anti-Moesin, Clone: MSN491, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328751ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328751M

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4478
Concentration 0.2mg/mL
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype κ
Quantity 1mL
Antigen Moesin
Immunogen Recombinant human Moesin protein
Gene Accession No. P26038
Test Specificity Recognizes 78kDa moesin protein. Moesin, a member of the talin-4.1 superfamily, is a linking protein of the sub-membranous actin cytoskeleton. It is expressed in variable amounts in cells of different phenotypes such as macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. The ERM proteins, ezrin, radixin, and moesin are involved in a variety of cellular functions, such as cell adhesion, migration, and the organization of cell surface structures, and are highly homologous, both in protein sequence and in functional activity, with merlin/schwannomin, a neurofibromatosis-2-associated tumor-suppressor protein. Cell lines of epithelial and mesothelial origin contain both moesin and radixin whereas cells of endothelial and lymphoid origin express moesin.
Clone MSN491
Conjugate Unlabeled
Cross Reactivity Human
Regulatory Status RUO
Format Purified
Host Species Mouse