Please login to your online account to display your discounted pricing

anti-MMP-2, Clone: 8B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200114

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200114

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Clone 8B4
Immunogen Activated recombinant human MMP2. [UniProt No. P08253]
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation PBS
Gene Accession No. P08253
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 4313
Concentration 1mg/mL
Conjugate Unlabeled
Antigen MMP-2
Regulatory Status RUO
Test Specificity This is specific for pro and active MMP2.
Quantity 0.1mL
Host Species Mouse
Format Purified