Please login to your online account to display your discounted pricing

anti-MMP-2, Clone: 8B4, Novus Biologicals™

$144.63 - $384.00

Specifications

Clone 8B4
Host Species Murine
Isotype IgG1, κ
Immunogen Activated recombinant human MMP2. [UniProt No. P08253]
Antigen MMP-2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB200114

 
novus biologicals canada
NB200114
Unlabeled 0.1mL Each for $384.00

NB200114SS

 
novus biologicals canada
NB200114SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Clone 8B4
Host Species Murine
Isotype IgG1, κ
Immunogen Activated recombinant human MMP2. [UniProt No. P08253]
Antigen MMP-2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This is specific for pro and active MMP2.
Gene Accession No. P08253
Gene ID (Entrez) 4313
Formulation PBS
Concentration 1mg/mL