Please login to your online account to display your discounted pricing

anti-MMP-2, Clone: 8B4, Novus Biologicals™


Manufacturer: novus biologicals canada  NB200114SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB200114SS

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.025mL
Regulatory Status RUO
Format Purified
Gene Accession No. P08253
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 4313
Concentration 1mg/mL
Conjugate Unlabeled
Host Species Mouse
Immunogen Activated recombinant human MMP2. [UniProt No. P08253]
Isotype IgG1, κ
Clone 8B4
Antigen MMP-2
Formulation PBS
Test Specificity This is specific for pro and active MMP2.