Please login to your online account to display your discounted pricing

anti-MMP-12, Biotin, Clone: 706F9.01, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0281B

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0281B

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4321
Immunogen Recombinant HME
Quantity 0.1mg
Format Ion Exchange Chromatography
Formulation Antibody in Tris-NaCl pH 8
Applications ELISA
Isotype IgG1
Monoclonal or Polyclonal Monoclonal
Clone 706F9.01
Conjugate Biotin
Antigen MMP-12
Gene Accession No. NP_002417
Host Species Mouse
Regulatory Status RUO
Primary or Secondary Primary
Cross Reactivity Human