Please login to your online account to display your discounted pricing

anti-MMP-12, Biotin, Clone: 706F9.01, Novus Biologicals™

$406.00 - $536.00

Specifications

Clone 706F9.01
Host Species Mouse
Isotype IgG1
Immunogen Recombinant HME
Antigen MMP-12
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

DDX0281B005

 
novus biologicals canada
DDX0281B005MG
Biotin 0.05mg Each for $406.00

DDX0281B

 
novus biologicals canada
DDX0281B
Biotin 0.1mg Each for $536.00
Description & Specifications

Specifications

Clone 706F9.01
Host Species Mouse
Isotype IgG1
Immunogen Recombinant HME
Antigen MMP-12
Monoclonal or Polyclonal Monoclonal
Format Ion Exchange Chromatography
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Gene Accession No. NP_002417
Gene ID (Entrez) 4321
Formulation Antibody in Tris-NaCl pH 8