Please login to your online account to display your discounted pricing

anti-MMP-1, Clone: 6A5, Novus Biologicals™

$144.63 - $384.00

Specifications

Clone 6A5
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human MMP1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P03956]
Antigen MMP-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222123

 
novus biologicals canada
NBP222123
Unlabeled 0.1mL Each for $384.00

NBP222123SS

 
novus biologicals canada
NBP222123SS
Unlabeled 0.025mL Each for $144.63
Description & Specifications

Specifications

Clone 6A5
Host Species Murine
Isotype IgG1
Immunogen Purified recombinant fragment of human MMP1 expressed in &italic_begin;E. coli&italic_end;. [UniProt No. P03956]
Antigen MMP-1
Monoclonal or Polyclonal Monoclonal
Format Precipitation
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity Human and Mouse. Other species have not been tested.
Gene Accession No. P03956
Gene ID (Entrez) 4312
Formulation PBS
Concentration 1.0mg/mL