Please login to your online account to display your discounted pricing

anti-MLH1, Clone: 164C819, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 164C819
Host Species Mouse
Isotype IgG1, κ
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 387-403 DSREQKLDAFLQPLSKP of human MLH1; GenBank no. NP_000240.1.
Antigen MLH1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056552T

 
novus biologicals canada
NB10056552SS
Unlabeled 0.025mg Each for $139.00

NB10056552

 
novus biologicals canada
NB10056552
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Clone 164C819
Host Species Mouse
Isotype IgG1, κ
Immunogen This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 387-403 DSREQKLDAFLQPLSKP of human MLH1; GenBank no. NP_000240.1.
Antigen MLH1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity In Molt-4, a 83 kDa band is observed. Reacts with Human. 93% sequence identity with mouse and canine proteins.
Gene Accession No. P40692
Gene ID (Entrez) 4292
Formulation PBS containing 0.05% BSA