Please login to your online account to display your discounted pricing

anti-Mitofusin 1, Clone: 11E9-1H12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171775

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171775

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human and Mouse. Rat reactivity reported in scientific liteRature (PMID: 23658809).
Formulation Tris-glycine, 150mM NaCl
Primary or Secondary Primary
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Mouse
Isotype κ
Clone 11E9-1H12
Antigen Mitofusin 1
Immunogen Mouse Mitofusin-1 [Swiss-Prot No. Q811U4]
Quantity 0.1mL
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 55669