Please login to your online account to display your discounted pricing

anti-Mitofusin 1, Clone: 11E9-1H12, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 11E9-1H12
Host Species Mouse
Isotype κ
Immunogen Mouse Mitofusin-1 [Swiss-Prot No. Q811U4]
Antigen Mitofusin 1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP171775

 
novus biologicals canada
NBP171775
Unlabeled 0.1mL Each for $329.00

NBP171775SS

 
novus biologicals canada
NBP171775SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 11E9-1H12
Host Species Mouse
Isotype κ
Immunogen Mouse Mitofusin-1 [Swiss-Prot No. Q811U4]
Antigen Mitofusin 1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Human and Mouse. Rat reactivity reported in scientific liteRature (PMID: 23658809).
Gene ID (Entrez) 55669
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL