Please login to your online account to display your discounted pricing

anti-Mitofusin 1, Clone: 11E9-1H12, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP171775SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP171775SS

  • / Each
 

Description & Specifications

Specifications

Antigen Mitofusin 1
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 11E9-1H12
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation Tris-glycine, 150mM NaCl
Host Species Murine
Immunogen Mouse Mitofusin-1 [Swiss-Prot No. Q811U4]
Isotype κ
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 55669
Test Specificity Human and Mouse. Rat reactivity reported in scientific liteRature (PMID: 23658809).
Target Species Human
Target Species Murine
Target Species Rat