Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: MTC754, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232874

 View all options for this product


For Research Use Only

Catalog No. NBP232874


Description & Specifications


Antigen Mitochondria
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Clone MTC754
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Mitochondrial fraction of HeLa cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Shows broad species reactivity.