Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: MTC754, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone MTC754
Host Species Mouse
Isotype κ
Immunogen Mitochondrial fraction of HeLa cells
Antigen Mitochondria
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP2328741M

 
novus biologicals canada
NBP2328741ML
Unlabeled 1mL Each for $429.00

NBP232874

 
novus biologicals canada
NBP232874
Unlabeled 0.5mL Each for $321.00
Description & Specifications

Specifications

Clone MTC754
Host Species Mouse
Isotype κ
Immunogen Mitochondrial fraction of HeLa cells
Antigen Mitochondria
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Shows broad species reactivity.
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL