Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: MTC754, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328741ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328741M

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Antigen Mitochondria
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Immunogen Mitochondrial fraction of HeLa cells
Isotype κ
Clone MTC754
Test Specificity Shows broad species reactivity.
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Quantity 1mL
Concentration 0.2mg/mL
Format Purified
Host Species Mouse