Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: MTC719, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232873

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232873

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Mitochondrial fraction of HeLa cells
Primary or Secondary Primary
Cross Reactivity Human
Isotype κ
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Clone MTC719
Concentration 0.2mg/mL
Conjugate Unlabeled
Format Purified
Test Specificity Shows broad species reactivity. Does not react with insects or bacteria.
Quantity 0.5mL
Host Species Mouse
Antigen Mitochondria