Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: MTC719, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone MTC719
Host Species Mouse
Isotype κ
Immunogen Mitochondrial fraction of HeLa cells
Antigen Mitochondria
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232873

 
novus biologicals canada
NBP232873
Unlabeled 0.5mL Each for $321.00

NBP2328731M

 
novus biologicals canada
NBP2328731ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone MTC719
Host Species Mouse
Isotype κ
Immunogen Mitochondrial fraction of HeLa cells
Antigen Mitochondria
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Shows broad species reactivity. Does not react with insects or bacteria.
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL