Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: MTC719, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328731ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328731M

  • / Each
 

Description & Specifications

Specifications

Test Specificity Shows broad species reactivity. Does not react with insects or bacteria.
Conjugate Unlabeled
Quantity 1mL
Monoclonal or Polyclonal Monoclonal
Concentration 0.2mg/mL
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Mitochondrial fraction of HeLa cells
Regulatory Status RUO
Clone MTC719
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Cross Reactivity Human
Format Purified
Isotype κ
Antigen Mitochondria
Host Species Mouse