Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: 113-1, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2329801ML

 View all options for this product


For Research Use Only

Catalog No. NBP2329801M


Description & Specifications


Antigen Mitochondria
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone 113-1
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Semi-purified mitochondrial preparation
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Human. Does not react with Mouse or Rat.