Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: 113-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232980

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232980

  • / Each
 

Description & Specifications

Specifications

Test Specificity Human. Does not react with Mouse or Rat.
Concentration 0.2mg/mL
Conjugate Unlabeled
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Isotype κ
Format Purified
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Immunogen Semi-purified mitochondrial preparation
Primary or Secondary Primary
Clone 113-1
Antigen Mitochondria
Regulatory Status RUO
Quantity 0.5mL