Please login to your online account to display your discounted pricing

anti-Mitochondria, Clone: 113-1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone 113-1
Host Species Mouse
Isotype κ
Immunogen Semi-purified mitochondrial preparation
Antigen Mitochondria
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232980

 
novus biologicals canada
NBP232980
Unlabeled 0.5mL Each for $321.00

NBP2329801M

 
novus biologicals canada
NBP2329801ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone 113-1
Host Species Mouse
Isotype κ
Immunogen Semi-purified mitochondrial preparation
Antigen Mitochondria
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity Human. Does not react with Mouse or Rat.
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL