Please login to your online account to display your discounted pricing

anti-MITF, Clone: 21D1418, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 21D1418
Host Species Murine
Isotype IgG1
Immunogen A synthetic peptide mapping to the 400-450 a.a. region of accession number O75030.
Antigen MITF
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056561

 
novus biologicals canada
NB10056561
Unlabeled 0.1mg Each for $329.00

NB10056561T

 
novus biologicals canada
NB10056561SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 21D1418
Host Species Murine
Isotype IgG1
Immunogen A synthetic peptide mapping to the 400-450 a.a. region of accession number O75030.
Antigen MITF
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Mitf produces, by alternative splicing, 10 different transcripts, all with introns, putatively encoding 10 different protein isoforms. Therefore, observed molecular weights may vary depending on MITF isoforms expressed. Users are encouraged to utilize the peptide immunogen sequence to gain further information about the isoforms recognized by the antibody. Reacts with Human, Mouse, Rat. Predicted cross-reactivity based on sequence identity: Dog (100%), Pig (100%).
Gene Accession No. O75030
Gene ID (Entrez) 4286
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL