Please login to your online account to display your discounted pricing

anti-MITF, Clone: 21D1418, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056561

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056561

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Regulatory Status RUO
Format Purified
Gene ID (Entrez) 4286
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL
Isotype IgG1
Clone 21D1418
Monoclonal or Polyclonal Monoclonal
Gene Accession No. O75030
Host Species Mouse
Immunogen A synthetic peptide mapping to the 400-450 a.a. region of accession number O75030.
Applications Western Blotting
Quantity 0.1mg
Antigen MITF
Primary or Secondary Primary
Conjugate Unlabeled
Test Specificity Mitf produces, by alternative splicing, 10 different transcripts, all with introns, putatively encoding 10 different protein isoforms. Therefore, observed molecular weights may vary depending on MITF isoforms expressed. Users are encouraged to utilize the peptide immunogen sequence to gain further information about the isoforms recognized by the antibody. Reacts with Human, Mouse, Rat. Predicted cross-reactivity based on sequence identity: Dog (100%), Pig (100%).