Please login to your online account to display your discounted pricing

anti-MGMT, Clone: MT 23.2, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100168

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100168

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blotting
Isotype IgG2b
Quantity 0.1mL
Antigen MGMT
Conjugate Unlabeled
Test Specificity This is specific for MGMT (ref 1). Human. This antibody does not work in mouse.
Gene Accession No. P16455
Primary or Secondary Primary
Format Ascites
Gene ID (Entrez) 4255
Cross Reactivity Human
Host Species Mouse
Clone MT 23.2
Immunogen Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from &italic_begin;E. coli&italic_end; [UniProt No. P16455]
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal