Please login to your online account to display your discounted pricing

anti-MGMT, Clone: MT 23.2, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone MT 23.2
Host Species Murine
Isotype IgG2b
Immunogen Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from &italic_begin;E. coli&italic_end; [UniProt No. P16455]
Antigen MGMT
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100168

 
novus biologicals canada
NB100168
Unlabeled 0.1mL Each for $329.00

NB100168SS

 
novus biologicals canada
NB100168SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone MT 23.2
Host Species Murine
Isotype IgG2b
Immunogen Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from &italic_begin;E. coli&italic_end; [UniProt No. P16455]
Antigen MGMT
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Test Specificity This is specific for MGMT (ref 1). Human. This antibody does not work in mouse.
Gene Accession No. P16455
Gene ID (Entrez) 4255