Please login to your online account to display your discounted pricing

anti-Melan-A/MART-1, Clone: A103, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329781ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329781M

  • / Each
 

Description & Specifications

Specifications

Antigen Melan-A/MART-1
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Clone A103
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 2315