Please login to your online account to display your discounted pricing

anti-Melan-A/MART-1, Clone: A103, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232978

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232978

  • / Each
 

Description & Specifications

Specifications

Concentration 0.2mg/mL
Primary or Secondary Primary
Gene ID (Entrez) 2315
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene Accession No. Q16655
Isotype κ
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Regulatory Status RUO
Antigen Melan-A/MART-1
Formulation PBS with 0.05% BSA.
Immunogen Recombinant hMART-1 protein
Quantity 0.5mL
Clone A103
Format Purified