Please login to your online account to display your discounted pricing

anti-Melan-A/MART-1, Clone: A103, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone A103
Host Species Mouse
Isotype κ
Immunogen Recombinant hMART-1 protein
Antigen Melan-A/MART-1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232978

 
novus biologicals canada
NBP232978
Unlabeled 0.5mL Each for $321.00

NBP2329781M

 
novus biologicals canada
NBP2329781ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone A103
Host Species Mouse
Isotype κ
Immunogen Recombinant hMART-1 protein
Antigen Melan-A/MART-1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Gene Accession No. Q16655
Gene ID (Entrez) 2315
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL