missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-MAVS, Clone: 58N3E1, Novus Biologicals™

Manufacturer:  Novus Biologicals Canada NBP222378SS

 View more versions of this product

Catalog No. NBP222378SS


 

Disclaimers:

For Research Use Only


Specifications Description & Specifications

Specifications

MAVS
58N3E1
Unlabeled
AAH44952.1
Murine
κ
RUO
Monoclonal
Human
Western Blot
0.5mg/mL
0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Purified
Within the range of amino acids 93-298 of human MAVS (isoform 1) were used as the immunogen for this antibody.
0.025mg
Primary
57506