Please login to your online account to display your discounted pricing

anti-MAVS, Clone: 58N3E1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222378

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222378

  • / Each
 

Description & Specifications

Specifications

Antigen MAVS
Applications Western Blot
Clone 58N3E1
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. AAH44952.1
Host Species Murine
Immunogen Within the range of amino acids 93-298 of human MAVS (isoform 1) were used as the immunogen for this antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 57506
Target Species Human