Please login to your online account to display your discounted pricing

anti-MAVS, Clone: 58N3B6, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222380SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222380SS

  • / Each
 

Description & Specifications

Specifications

Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Isotype κ
Quantity 0.025mg
Immunogen Amino acids 93-276 of human MAVS (isoform 1) were used as the immunogen for this antibody.
Cross Reactivity Human
Format Purified
Host Species Mouse
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen MAVS
Clone 58N3B6
Gene ID (Entrez) 57506
Gene Accession No. AAH44952.1
Primary or Secondary Primary
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Conjugate Unlabeled