Please login to your online account to display your discounted pricing

anti-MAVS, Clone: 58N2B2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222379SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222379SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 57506
Primary or Secondary Primary
Conjugate Unlabeled
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. AAH44952.1
Monoclonal or Polyclonal Monoclonal
Immunogen MAVS(IPS-1)
Isotype κ
Cross Reactivity Human
Quantity 0.025mg
Format Purified
Host Species Mouse
Applications Western Blotting
Antigen MAVS
Clone 58N2B2
Regulatory Status RUO