Please login to your online account to display your discounted pricing

anti-MAVS, Clone: 58N2B2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222379

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222379

  • / Each
 

Description & Specifications

Specifications

Antigen MAVS
Applications Western Blot
Clone 58N2B2
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. AAH44952.1
Host Species Murine
Immunogen MAVS(IPS-1)
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 57506
Target Species Human