Please login to your online account to display your discounted pricing

anti-MAVS, Clone: 58N2B2, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 58N2B2
Host Species Murine
Isotype κ
Immunogen MAVS(IPS-1)
Antigen MAVS
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222379SS

 
novus biologicals canada
NBP222379SS
Unlabeled 0.025mg Each for $139.00

NBP222379

 
novus biologicals canada
NBP222379
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Clone 58N2B2
Host Species Murine
Isotype κ
Immunogen MAVS(IPS-1)
Antigen MAVS
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. AAH44952.1
Gene ID (Entrez) 57506
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA