Please login to your online account to display your discounted pricing

anti-Lung Specific Antigen, Clone: LSA120, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP232870

 View all options for this product


For Research Use Only

Catalog No. NBP232870


Description & Specifications


Antigen Lung Specific Antigen
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Clone LSA120
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Protein extract of human lung cells
Isotype κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal