Please login to your online account to display your discounted pricing

anti-Lung Specific Antigen, Clone: LSA120, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LSA120
Host Species Mouse
Isotype κ
Immunogen Protein extract of human lung cells
Antigen Lung Specific Antigen
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232870

 
novus biologicals canada
NBP232870
Unlabeled 0.5mL Each for $321.00

NBP2328701M

 
novus biologicals canada
NBP2328701ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone LSA120
Host Species Mouse
Isotype κ
Immunogen Protein extract of human lung cells
Antigen Lung Specific Antigen
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL