Please login to your online account to display your discounted pricing

anti-Lung Specific Antigen, Clone: LSA120, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2328701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2328701M

  • / Each
 

Description & Specifications

Specifications

Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Cross Reactivity Human
Isotype κ
Host Species Mouse
Regulatory Status RUO
Quantity 1mL
Clone LSA120
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL
Format Purified
Immunogen Protein extract of human lung cells
Antigen Lung Specific Antigen