Please login to your online account to display your discounted pricing

anti-LDL R, Clone: C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178159SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178159SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Quantity 0.025mL
Clone C7
Immunogen Partially purified bovine LDL receptor [Swiss-Prot No. P01131]
Host Species Mouse
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Radioimmunoassay
Applications Western Blotting
Primary or Secondary Primary
Conjugate Unlabeled
Concentration 1mg/mL
Isotype IgG2b
Format Purified
Antigen LDL R
Gene ID (Entrez) 3949
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P01130