Please login to your online account to display your discounted pricing

anti-LDL R, Clone: C7, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178159

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178159

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mL
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Radioimmunoassay
Applications Western Blotting
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Porcine
Immunogen Partially purified bovine LDL receptor [Swiss-Prot No. P01131]
Antigen LDL R
Regulatory Status RUO
Clone C7
Format Purified
Isotype IgG2b
Formulation Tris-glycine, 150mM NaCl
Gene Accession No. P01130
Monoclonal or Polyclonal Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled
Gene ID (Entrez) 3949
Primary or Secondary Primary
Host Species Mouse