Please login to your online account to display your discounted pricing

anti-Lck, Clone: 33D196, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 33D196
Host Species Murine
Isotype IgG1
Immunogen This monoclonal antibody was raised against a synthetic peptide containing amino acid residues of human lck protein.
Antigen Lck
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056678

 
novus biologicals canada
NB10056678
Unlabeled 0.2mL Each for $329.00

NB10056678T

 
novus biologicals canada
NB10056678SS
Unlabeled 0.05mL Each for $139.00
Description & Specifications

Specifications

Clone 33D196
Host Species Murine
Isotype IgG1
Immunogen This monoclonal antibody was raised against a synthetic peptide containing amino acid residues of human lck protein.
Antigen Lck
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity Lck (33D196).
Gene Accession No. P06239
Gene ID (Entrez) 3932
Formulation PBS
Concentration 0.5mg/mL