Please login to your online account to display your discounted pricing

anti-LANA2, Clone: CM-A807, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone CM-A807
Host Species Mouse
Isotype IgG1, κ
Immunogen Recombinant LANA2 fusion protein.
Antigen LANA2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB200167

 
novus biologicals canada
NB200167
Unlabeled 0.1mL Each for $309.05

NB200167SS

 
novus biologicals canada
NB200167SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone CM-A807
Host Species Mouse
Isotype IgG1, κ
Immunogen Recombinant LANA2 fusion protein.
Antigen LANA2
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This is specific for KSHV LANA2 (residues 143-310 epitope recognition). Reacts with viral LANA2 protein and shows no cross-reactivity to cellular proteins. Also reacts with human and mouse viral KSHV proteins.
Formulation PBS
Concentration 2mg/mL