Please login to your online account to display your discounted pricing

anti-LAMP-1/CD107a, Clone: 6E2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225155SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225155SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 3916
Gene Accession No. P11279
Test Specificity Human, Bovine, Porcine, Mouse and Rat. Other species have not been tested.
Concentration 1.6mg/mL
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Isotype IgG1
Immunogen Full length human LAMP1 [UniProt No. P11279]
Regulatory Status RUO
Quantity 0.025mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Formulation PBS
Format Affinity Purified
Host Species Mouse
Antigen LAMP-1/CD107a
Clone 6.00E+02